siRNA Details
| Gene Symbol: Tssk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253888.1 | NP_001240817.1 | 4 | 1214 | ||
| NM_001253889.1 | NP_001240818.1 | 4 | 1361 | |||
| NM_027673.3 | NP_081949.1 | 4 | 1184 | |||
| GenBank mRNA | AK006144.1 | - | 4 | 1336 | ||
| AK016890.1 | - | 4 | 1186 | |||
| AY461664.1 | - | 4 | 1076 | |||
| EF127819.1 | - | 4 | 1186 | |||
| EF127820.1 | - | 4 | 1216 | |||
| EF127821.1 | - | 4 | 1362 | |||
| EF127822.1 | - | 3 | 1629 | |||

Tssk4
Genomic Map