siRNA Details
| Gene Symbol: Lrrc72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027699.2 | NP_081975.1 | 5 | 420 | ||
| GenBank mRNA | AK007140.1 | - | 5 | 421 | ||
| BC107290.1 | - | 4 | 326 | 40053043 | ||
| BC107291.1 | - | 4 | 326 | 40053045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc72| Gene Symbol: Lrrc72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027699.2 | NP_081975.1 | 5 | 420 | ||
| GenBank mRNA | AK007140.1 | - | 5 | 421 | ||
| BC107290.1 | - | 4 | 326 | 40053043 | ||
| BC107291.1 | - | 4 | 326 | 40053045 | ||