siRNA Details
Gene Symbol: 4933421I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027702.2 | NP_081978.1 | 4 | 1132 | ||
GenBank mRNA | AK016864.1 | - | 4 | 1133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933421I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027702.2 | NP_081978.1 | 4 | 1132 | ||
GenBank mRNA | AK016864.1 | - | 4 | 1133 |