siRNA Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163661.1 | NP_001157133.1 | 3 | 1168 | ||
| NM_027880.2 | NP_082156.2 | 3 | 1079 | |||
| GenBank mRNA | AK004582.1 | - | 3 | 949 | ||
| AK028658.1 | - | 3 | 933 | |||
| AK050773.1 | - | 3 | 899 | |||
| AK084830.1 | - | 3 | 949 | |||
| AK129189.1 | - | 3 | 1117 | |||
| AK143827.1 | - | 3 | 1169 | |||
| AK146963.1 | - | 3 | 955 | |||
| AK154727.1 | - | 3 | 981 | |||
| AK180072.1 | - | 2 | 348 | |||
| AK181385.1 | - | 1 | 83 | |||
| AK182669.1 | - | 1 | 83 | |||
| BC011077.1 | - | 3 | 929 | 3984911 | ||
| BC080713.1 | - | 3 | 1108 | |||

Telo2
Genomic Map