siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB093224.1 | - | 5 | 1403 | ||
| AF363690.1 | - | 10 | 2133 | |||
| AK147276.1 | - | 10 | 2766 | |||
| AK147535.1 | - | 11 | 4884 | |||
| BC058665.1 | - | 7 | 1651 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cic| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB093224.1 | - | 5 | 1403 | ||
| AF363690.1 | - | 10 | 2133 | |||
| AK147276.1 | - | 10 | 2766 | |||
| AK147535.1 | - | 11 | 4884 | |||
| BC058665.1 | - | 7 | 1651 | |||