siRNA Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027886.3 | NP_082162.3 | 25 | 3600 | ||
| GenBank mRNA | AK004757.2 | - | 25 | 3553 | ||
| AK156122.1 | - | 24 | 3688 | |||
| AK165911.1 | - | 25 | 3592 | |||
| AK171989.1 | - | 24 | 3462 | |||
| AK209763.1 | - | 1 | 281 | |||
| BC035530.1 | - | 11 | 2640 | |||
| BC042403.1 | - | 23 | 3637 | |||
| BC061635.1 | - | 25 | 3583 | 6530178 | ||

Stk11ip
Genomic Map