siRNA Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027901.2 | NP_082177.2 | 19 | 3316 | ||
| GenBank mRNA | AK004895.1 | - | 18 | 2623 | ||
| AK008582.1 | - | 2 | 629 | |||
| AK012095.1 | - | 12 | 1913 | |||
| AK083585.1 | - | 19 | 3319 | |||
| AK083636.1 | - | 18 | 3071 | |||
| AK129033.1 | - | 18 | 3066 | |||
| AK149771.1 | - | 19 | 2742 | |||
| AK151300.1 | - | 19 | 3081 | |||
| AK152439.1 | - | 19 | 3081 | |||
| AK154056.1 | - | 6 | 1091 | |||
| AK159938.1 | - | 19 | 3084 | |||
| BC026573.1 | - | 11 | 1634 | |||
| BC034369.1 | - | 19 | 3094 | 3994677 | ||
| BC043100.1 | - | 18 | 3482 | |||

Gtf3c2
Genomic Map