siRNA Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027903.3 | NP_082179.1 | 7 | 1639 | ||
| GenBank mRNA | AK005050.2 | - | 7 | 1640 | ||
| AK154452.1 | - | 7 | 1621 | |||
| BC031710.1 | - | 7 | 1617 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhdh| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027903.3 | NP_082179.1 | 7 | 1639 | ||
| GenBank mRNA | AK005050.2 | - | 7 | 1640 | ||
| AK154452.1 | - | 7 | 1621 | |||
| BC031710.1 | - | 7 | 1617 | |||