siRNA Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027924.2 | NP_082200.1 | 1, 2 | 307 | ||
| GenBank mRNA | AF335583.1 | - | 1, 2 | 124 | ||
| AK003359.2 | - | 1, 2 | 309 | |||
| BC030896.1 | - | 1, 2 | 258 | 4489485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfd| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027924.2 | NP_082200.1 | 1, 2 | 307 | ||
| GenBank mRNA | AF335583.1 | - | 1, 2 | 124 | ||
| AK003359.2 | - | 1, 2 | 309 | |||
| BC030896.1 | - | 1, 2 | 258 | 4489485 | ||