siRNA Details
| Gene Symbol: Zbtb43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025594.1 | NP_001020765.1 | 3 | 327 | ||
| NM_027947.2 | NP_082223.2 | 4 | 419 | |||
| GenBank mRNA | AK005818.1 | - | 4 | 393 | ||
| AK122277.1 | - | 3 | 298 | |||
| AK147806.1 | - | 3 | 322 | |||
| BC043117.1 | - | 3 | 327 | 6408745 | ||

Zbtb43
Genomic Map