siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027992.3 | NP_082268.2 | 3 | 518 | ||
| GenBank mRNA | AK009646.1 | - | 3 | 421 | ||
| AK018015.1 | - | 3 | 425 | |||
| AK148477.1 | - | 3 | 440 | |||
| BC050246.1 | - | 3 | 435 | 6313498 | ||

Tmem106b
Genomic Map