siRNA Details
| Gene Symbol: Myh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271538.1 | NP_001258467.1 | 1 | 320 | ||
| NM_001271540.1 | NP_001258469.1 | 2 | 361 | |||
| NM_028021.3 | NP_082297.1 | 1 | 320 | |||
| GenBank mRNA | AY205605.1 | - | 1 | 320 | ||
| AY363100.1 | - | 2 | 361 | |||
| EF602040.1 | - | 1 | 320 | |||

Myh14
Genomic Map