siRNA Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028024.2 | NP_082300.1 | 3 | 193 | ||
| GenBank mRNA | AK010371.1 | - | 3 | 193 | ||
| AK015918.1 | - | 198 | ||||
| AK052645.1 | - | 3 | 193 | |||
| AK083475.1 | - | 3 | 236 | |||
| AK149696.1 | - | 3 | 337 | |||
| AK169878.1 | - | 3 | 218 | |||
| BC013469.1 | - | 2 | 146 | 4219892 | ||
| BC024398.1 | - | 2 | 136 | 5003001 | ||

Nkiras2
Genomic Map