siRNA Details
| Gene Symbol: Slc25a35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028048.2 | NP_082324.1 | 2 | 939 | ||
| GenBank mRNA | AK007453.1 | - | 2 | 941 | ||
| AK035739.1 | - | 922 | ||||
| AK157873.1 | - | 2 | 933 | |||
| AK171131.1 | - | 2 | 946 | |||
| BC004569.1 | - | 2 | 379 | |||
| BC019996.1 | - | 2 | 881 | 4207271 | ||

Slc25a35
Genomic Map