siRNA Details
| Gene Symbol: Slc39a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136237.1 | NP_001129709.1 | 3 | 325 | ||
| NM_028051.3 | NP_082327.2 | 2 | 411 | |||
| NM_028092.3 | NP_082368.1 | 2 | 287 | |||
| GenBank mRNA | AK007473.1 | - | 2 | 412 | ||
| AK008448.1 | - | 2 | 288 | |||
| BC028990.1 | - | 3 | 286 | 5041383 | ||
| CA492656.1 | - | 3 | 365 | |||

Slc39a5
Genomic Map