siRNA Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028053.2 | NP_082329.1 | 2 | 256 | ||
| GenBank mRNA | AB261159.1 | - | 2 | 192 | ||
| AK005479.1 | - | 2 | 239 | |||
| AK150377.1 | - | 2 | 234 | |||
| AK150892.1 | - | 2 | 234 | |||
| AK153075.1 | - | 2 | 221 | |||
| BC011072.1 | - | 2 | 234 | 3978742 | ||
| BY138283.1 | - | 2 | 256 | |||

Tmem38b
Genomic Map