siRNA Details
| Gene Symbol: Slc39a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028064.2 | NP_082340.1 | 4 | 810 | ||
| GenBank mRNA | AK005535.1 | - | 4 | 850 | ||
| AK146977.1 | - | 4 | 851 | |||
| AK147107.1 | - | 4 | 829 | |||
| BC023498.1 | - | 4 | 810 | 5011088 | ||

Slc39a4
Genomic Map