siRNA Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF356627.1 | - | 3 | 186 | ||
| AK005546.1 | - | 3 | 198 | |||
| BC019485.1 | - | 3 | 175 | 4194672 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF356627.1 | - | 3 | 186 | ||
| AK005546.1 | - | 3 | 198 | |||
| BC019485.1 | - | 3 | 175 | 4194672 | ||