siRNA Details
| Gene Symbol: 1810055G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028077.2 | NP_082353.1 | 4 | 928 | ||
| GenBank mRNA | AK007868.1 | - | 4 | 929 | ||
| AK154828.1 | - | 4 | 1250 | |||
| BC019471.1 | - | 4 | 922 | 4161022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810055G02Rik| Gene Symbol: 1810055G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028077.2 | NP_082353.1 | 4 | 928 | ||
| GenBank mRNA | AK007868.1 | - | 4 | 929 | ||
| AK154828.1 | - | 4 | 1250 | |||
| BC019471.1 | - | 4 | 922 | 4161022 | ||