siRNA Details
| Gene Symbol: Seh1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039088.1 | NP_001034177.1 | 4 | 459 | ||
| NM_028112.2 | NP_082388.1 | 4 | 459 | |||
| GenBank mRNA | AK011330.1 | - | 4 | 361 | ||
| AK088835.1 | - | 4 | 333 | |||
| AK143727.1 | - | 4 | 459 | |||
| AK166573.1 | - | 4 | 324 | |||
| AK169231.1 | - | 4 | 340 | |||
| AK169388.1 | - | 4 | 339 | |||
| AK170591.1 | - | 4 | 412 | |||
| BC027244.1 | - | 4 | 411 | 3584697 | ||
| BY732540.1 | - | 4 | 459 | |||
| Gene Symbol: 4930549G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CN666923.1 | - | 1 | 20 | ||

Seh1l
Genomic Map