siRNA Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027924.2 | NP_082200.1 | 7 | 1338 | ||
| GenBank mRNA | AK003359.2 | - | 7 | 1340 | ||
| AK141551.1 | - | 7 | 1349 | |||
| BC030896.1 | - | 7 | 1074 | 4489485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfd| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027924.2 | NP_082200.1 | 7 | 1338 | ||
| GenBank mRNA | AK003359.2 | - | 7 | 1340 | ||
| AK141551.1 | - | 7 | 1349 | |||
| BC030896.1 | - | 7 | 1074 | 4489485 | ||