siRNA Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028018.2 | NP_082294.1 | 10 | 1202 | ||
| GenBank mRNA | AK010262.1 | - | 10 | 1172 | ||
| AK050468.1 | - | 9 | 2502 | |||
| AK088192.1 | - | 10 | 1157 | |||
| AK090233.1 | - | 1 | 210 | |||
| AK122215.1 | - | 5 | 5673 | |||
| AK152830.1 | - | 10 | 1169 | |||
| AK188961.1 | - | 1 | 20 | |||
| AK190075.1 | - | 1 | 20 | |||
| AK210859.1 | - | 1 | 116 | |||
| AK213979.1 | - | 1 | 103 | |||
| AK215893.1 | - | 1 | 20 | |||
| BC017605.1 | - | 10 | 1168 | 4507347 | ||
| BC039052.1 | - | 9 | 2478 | 2631157 | ||

Ist1
Genomic Map