siRNA Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028024.2 | NP_082300.1 | 4 | 1965 | ||
| GenBank mRNA | AK010371.1 | - | 4 | 1965 | ||
| AK052645.1 | - | 4 | 1965 | |||
| AK083475.1 | - | 4 | 2008 | |||
| AK169878.1 | - | 4 | 1990 | |||
| AK197962.1 | - | 1 | 161 | |||
| AK201543.1 | - | 1 | 161 | |||
| AK205184.1 | - | 1 | 161 | |||
| BC013469.1 | - | 3 | 1918 | 4219892 | ||
| BC024398.1 | - | 3 | 1908 | 5003001 | ||

Nkiras2
Genomic Map