siRNA Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028047.2 | NP_082323.1 | 14 | 2203 | ||
| GenBank mRNA | AK005126.1 | - | 14 | 2203 | ||
| AK039368.1 | - | 14 | 2202 | |||
| AK141576.1 | - | 14 | 2194 | |||
| AK169651.1 | - | 14 | 2127 | |||
| AK194093.1 | - | 1 | 232 | |||
| BC034211.1 | - | 14 | 2118 | 4988361 | ||

Smg9
Genomic Map