siRNA Details
| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028049.2 | NP_082325.2 | 7 | 1507 | ||
| GenBank mRNA | AK002745.1 | - | 7 | 1387 | ||
| AK005473.1 | - | 6 | 1335 | |||
| BC018273.1 | - | 7 | 1447 | 4458296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo22| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028049.2 | NP_082325.2 | 7 | 1507 | ||
| GenBank mRNA | AK002745.1 | - | 7 | 1387 | ||
| AK005473.1 | - | 6 | 1335 | |||
| BC018273.1 | - | 7 | 1447 | 4458296 | ||