siRNA Details
| Gene Symbol: Slc39a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136237.1 | NP_001129709.1 | 8 | 1185 | ||
| NM_028051.3 | NP_082327.2 | 7 | 1271 | |||
| NM_028092.3 | NP_082368.1 | 7 | 1147 | |||
| GenBank mRNA | AK007473.1 | - | 7 | 1272 | ||
| AK008448.1 | - | 7 | 1148 | |||
| BC028990.1 | - | 8 | 1146 | 5041383 | ||

Slc39a5
Genomic Map