siRNA Details
| Gene Symbol: Tmem68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028097.3 | NP_082373.1 | 4 | 550 | ||
| GenBank mRNA | AK008493.1 | - | 4 | 584 | ||
| AK048210.1 | - | 4 | 550 | |||
| AK084881.1 | - | 4 | 584 | |||
| AK088600.1 | - | 4 | 593 | |||
| AK147765.1 | - | 4 | 583 | |||
| AK162563.1 | - | 4 | 583 | |||
| AK165916.1 | - | 4 | 612 | |||
| AK166237.1 | - | 4 | 636 | |||
| BC016240.1 | - | 4 | 568 | 4506688 | ||

Tmem68
Genomic Map