siRNA Details
| Gene Symbol: MUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000255.3 | NP_000246.2 | 8 | 1811 | ||
| XM_005249143.3 | XP_005249200.1 | 8 | 1716 | |||
| GenBank mRNA | AB208845.1 | - | 3 | 389 | ||
| AK292568.1 | - | 8 | 1787 | |||
| AK312611.1 | - | 8 | 1786 | |||
| BC016282.1 | - | 8 | 1674 | 3908548 | ||
| BT007434.1 | - | 7 | 1546 | |||
| BX647789.1 | - | 8 | 1636 | |||
| KF030882.1 | - | 7 | 1546 | |||
| KU178201.1 | - | 7 | 1546 | |||
| KU178202.1 | - | 7 | 1546 | |||
| M65131.1 | - | 8 | 1622 | |||

MUT
Genomic Map