siRNA Details
| Gene Symbol: Zfp157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028130.3 | NP_082406.2 | 3 | 323 | ||
| GenBank mRNA | AI789072.1 | - | 3 | 323 | ||
| AK028290.1 | - | 3 | 283 | |||
| AK042470.1 | - | 2 | 2956 | |||
| BC049676.1 | - | 3 | 288 | |||
| BC059261.1 | - | 3 | 295 | 6409233 | ||

Zfp157
Genomic Map