siRNA Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028360.2 | NP_082636.3 | 3 | 420 | ||
| NM_029704.2 | NP_083980.2 | 3 | 420 | |||
| GenBank mRNA | AK013368.1 | - | 3 | 412 | ||
| AK034075.1 | - | 3 | 410 | |||
| BC026866.1 | - | 1 | 64 | |||
| BC037111.1 | - | 2 | 144 | 5370464 | ||

Ttc19
Genomic Map