siRNA Details
| Gene Symbol: Strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252448.1 | NP_001239377.1 | 6 | 430 | ||
| NM_001252449.1 | NP_001239378.1 | 4 | 319 | |||
| NM_028126.3 | NP_082402.1 | 4 | 319 | |||
| GenBank mRNA | AK011451.1 | - | 4 | 265 | ||
| AK138358.1 | - | 6 | 431 | |||
| AY341872.1 | - | 4 | 278 | |||
| AY341878.1 | - | 4 | 278 | |||
| BC052063.1 | - | 450 | ||||
| BC058517.1 | - | 4 | 220 | 5717255 | ||

Strada
Genomic Map