siRNA Details
| Gene Symbol: Psca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028216.2 | NP_082492.1 | 3 | 828 | ||
| GenBank mRNA | AF319173.1 | - | 3 | 816 | ||
| AK008851.1 | - | 3 | 829 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psca| Gene Symbol: Psca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028216.2 | NP_082492.1 | 3 | 828 | ||
| GenBank mRNA | AF319173.1 | - | 3 | 816 | ||
| AK008851.1 | - | 3 | 829 | |||