siRNA Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028351.3 | NP_082627.3 | 2 | 747 | ||
| GenBank mRNA | AB055811.1 | - | 2 | 705 | ||
| AK013366.1 | - | 2 | 696 | |||
| AK076308.1 | - | 694 | ||||
| AK133916.1 | - | 2 | 748 | |||
| AY864332.1 | - | 2 | 208 | |||
| BC103794.1 | - | 2 | 681 | 6517974 | ||
| BC145929.1 | - | 2 | 352 | 40131232 | ||
| BC145931.1 | - | 2 | 352 | 40131234 | ||

Rspo3
Genomic Map