siRNA Details
| Gene Symbol: Zfp157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028130.3 | NP_082406.2 | 5 | 1810 | ||
| GenBank mRNA | AK011480.1 | - | 1 | 400 | ||
| AK028290.1 | - | 5 | 1771 | |||
| BC059261.1 | - | 5 | 1782 | 6409233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp157| Gene Symbol: Zfp157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028130.3 | NP_082406.2 | 5 | 1810 | ||
| GenBank mRNA | AK011480.1 | - | 1 | 400 | ||
| AK028290.1 | - | 5 | 1771 | |||
| BC059261.1 | - | 5 | 1782 | 6409233 | ||