siRNA Details
| Gene Symbol: Zfp661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111029.1 | NP_001104499.1 | 5 | 2124 | ||
| NM_028141.3 | NP_082417.3 | 6 | 2341 | |||
| GenBank mRNA | AK013000.1 | - | 1 | 1236 | ||
| AK033122.1 | - | 5 | 2126 | |||
| AK134651.1 | - | 6 | 3395 | |||
| BC052876.1 | - | 6 | 2345 | 30047253 | ||

Zfp661
Genomic Map