siRNA Details
| Gene Symbol: 1600014C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085385.1 | NP_001078854.1 | 3 | 1262 | ||
| NM_028166.3 | NP_082442.1 | 3 | 1070 | |||
| GenBank mRNA | AK032135.1 | - | 3 | 1039 | ||
| AK032342.1 | - | 3 | 1041 | |||
| AK146474.1 | - | 3 | 1051 | |||
| BC085480.1 | - | 3 | 1008 | 6401538 | ||

1600014C10Rik
Genomic Map