siRNA Details
| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF079972.1 | - | 12 | 1135 | ||
| BC018505.1 | - | 11 | 1028 | |||
| BC139056.1 | - | 12 | 1177 | 8862078 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyth4| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF079972.1 | - | 12 | 1135 | ||
| BC018505.1 | - | 11 | 1028 | |||
| BC139056.1 | - | 12 | 1177 | 8862078 | ||