siRNA Details
| Gene Symbol: Plxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163608.1 | NP_001157080.1 | 3 | 420 | ||
| NM_028199.3 | NP_082475.3 | 2, 3 | 399 | |||
| GenBank mRNA | AF378760.1 | - | 2, 3 | 333 | ||
| AK010361.1 | - | 2, 3 | 399 | |||
| AK036144.1 | - | 3 | 374 | |||
| AK040620.1 | - | 2, 3 | 387 | |||
| AK153684.1 | - | 2, 3 | 386 | |||
| BC114352.2 | - | 2, 3 | 319 | 40057410 | ||

Plxdc1
Genomic Map