siRNA Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163531.1 | NP_001157003.1 | 11 | 1064 | ||
| NM_001163532.1 | NP_001157004.1 | 11 | 1067 | |||
| NM_028223.3 | NP_082499.3 | 11 | 1174 | |||
| GenBank mRNA | AK013879.1 | - | 11 | 997 | ||
| AK034579.1 | - | 11 | 1043 | |||
| AK082765.1 | - | 11 | 1035 | |||
| AK170273.1 | - | 11 | 1017 | |||
| BC005542.1 | - | 11 | 982 | 3590497 | ||

Tmem175
Genomic Map