siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011832.1 | - | 3, 4 | 279 | ||
| AK030398.1 | - | 3, 4 | 478 | |||
| AK047429.1 | - | 2, 3 | 261 | |||
| AK135723.1 | - | 2, 3 | 262 | |||
| AK162903.1 | - | 2, 3 | 701 | |||
| AY092766.1 | - | 2, 3 | 370 | |||
| BC023762.1 | - | 3, 4 | 548 | |||
| BC057991.1 | - | 3, 4 | 319 | 5323559 | ||
| BE994245.1 | - | 2, 3 | 420 | |||
| EF410128.1 | - | 503 | ||||

Zfp131
Genomic Map