siRNA Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028250.3 | NP_082526.2 | 4, 5 | 993 | ||
| GenBank mRNA | AK011781.1 | - | 4, 5 | 986 | ||
| AK167959.1 | - | 5, 6 | 933 | |||
| AK209754.1 | - | 4, 5 | 308 | |||
| AK213898.1 | - | 1, 2 | 64 | |||
| BC115580.1 | - | 4, 5 | 585 | 40102241 | ||
| BC115581.1 | - | 4, 5 | 585 | 40102242 | ||

Acbd6
Genomic Map