siRNA Details
| Gene Symbol: 2700097O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028314.2 | NP_082590.2 | 3 | 342 | ||
| GenBank mRNA | AK012621.1 | - | 3 | 342 | ||
| AK079348.1 | - | 3 | 341 | |||
| BC056967.1 | - | 3 | 336 | 5694212 | ||
| CX231570.1 | - | 3 | 408 | |||

2700097O09Rik
Genomic Map