siRNA Details
| Gene Symbol: 2810032G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012851.1 | - | 1226 | |||
| AK028030.1 | - | 1110 | ||||
| AK029678.1 | - | 1348 | ||||
| BC065777.1 | - | 590 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_028318.1 | - | 1348 | Chr. - - [-] on Build | ||

2810032G03Rik
Genomic Map