siRNA Details
| Gene Symbol: Mon1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028369.3 | NP_082645.1 | 4 | 1000 | ||
| GenBank mRNA | AK013387.1 | - | 4 | 947 | ||
| AK172624.1 | - | 4 | 907 | |||
| BC048852.1 | - | 4 | 926 | 5148425 | ||
| BC058717.1 | - | 4 | 978 | 5704178 | ||

Mon1a
Genomic Map