siRNA Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028472.2 | NP_082748.1 | 2 | 738 | ||
| GenBank mRNA | AF454954.1 | - | 2 | 738 | ||
| AK049718.1 | - | 2 | 530 | |||
| AY263358.1 | - | 2 | 259 | |||
| BC066153.1 | - | 2 | 243 | 30098453 | ||

Bmper
Genomic Map