siRNA Details
| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028550.3 | NP_082826.1 | 6, 7 | 973 | ||
| GenBank mRNA | AK006951.1 | - | 5, 6 | 847 | ||
| AK030264.1 | - | 6, 7 | 973 | |||
| BC061035.1 | - | 5, 6 | 884 | 6774253 | ||

1700074P13Rik
Genomic Map