siRNA Details
| Gene Symbol: 1700080E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028562.3 | NP_082838.2 | 2 | 375 | ||
| GenBank mRNA | AK006953.1 | - | 2 | 376 | ||
| BC048436.1 | - | 2 | 364 | 6773931 | ||
| FJ211595.1 | - | 2 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700080E11Rik| Gene Symbol: 1700080E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028562.3 | NP_082838.2 | 2 | 375 | ||
| GenBank mRNA | AK006953.1 | - | 2 | 376 | ||
| BC048436.1 | - | 2 | 364 | 6773931 | ||
| FJ211595.1 | - | 2 | 221 | |||