siRNA Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028617.2 | NP_082893.1 | 3 | 244 | ||
| GenBank mRNA | AK003644.1 | - | 3 | 224 | ||
| BC020119.1 | - | 3 | 244 | 3982577 | ||
| BC027419.1 | - | 3 | 220 | 4163554 | ||
| BC087734.1 | - | 3 | 241 | 5101734 | ||
| BC096439.1 | - | 2 | 401 | |||

Mvb12a
Genomic Map