siRNA Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028627.2 | NP_082903.2 | 18 | 3631 | ||
| GenBank mRNA | AK003542.1 | - | 2 | 607 | ||
| AK158685.1 | - | 8 | 1439 | |||
| AK158851.1 | - | 8 | 1563 | |||
| AK220438.1 | - | 13 | 3749 | |||
| BC043135.1 | - | 14 | 2461 | |||
| BC057353.1 | - | 18 | 3633 | |||
| BC058352.1 | - | 4 | 1006 | |||
| BC069847.1 | - | 1 | 328 | |||

Psd
Genomic Map