siRNA Details
| Gene Symbol: Arsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028710.3 | NP_082986.3 | 4 | 1048 | ||
| GenBank mRNA | AK018132.2 | - | 1 | 465 | ||
| AK158726.1 | - | 4 | 1049 | |||
| AK173082.1 | - | 1 | 147 | |||
| BC084731.1 | - | 1 | 738 | 6813261 | ||

Arsg
Genomic Map